Learn More
Gemini with LowDoseID, S5, Gray Boot
Catalog No. 17103071 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Handheld Chemical Analyzer
Catalog No. Product Type
17-103-071 Handheld Chemical Analyzer
1 options
Catalog No. 17-103-071 Supplier Thermo Scientific Supplier No. 8000347101
Add to Cart
Add to Cart
Request a Quote
Rugged handheld chemical analyzer with integrating Raman and FTIR spectroscopy for the identification of unknown solid and liquid chemicals and explosives.
Including LowDoseID, Screener software, 5-year warranty, 5-year support, and on-site training within the continental United States (CONUS; One 4-hour course per instrument for up to 12 students. Expires 9 months after date of purchase).
TRUSTED_SUSTAINABILITY
Specifications
Product Type Handheld Chemical Analyzer
Display Digital