Learn More
Stoelting™ Gemini-Mixter Hemostats
SDP
Catalog No. 10000478 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Full-curved; Delicate
Catalog No. Model
10-000-478 Full-curved; Delicate
1 options
Catalog No. 10-000-478 Supplier Stoelting™ Supplier No. 5211031P
Add to Cart
Add to Cart
Stoelting™ Gemini-Mixter Hemostats are used in many surgical procedures usually to control bleeding or the flow of liquids in tubing.
  • Feature scissor-like rring handles
  • Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure
Specifications
For Use With (Application) Neuroscience
Length (English) 5.5 in.
Model Full-curved; Delicate