Learn More
Gemini Test Kit
Catalog No. 17103070 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Gemini Chemical Identification Analyzer
Catalog No. For Use With (Equipment)
17-103-070 Gemini Chemical Identification Analyzer
1 options
Catalog No. 17-103-070 Supplier Thermo Scientific Supplier No. 8100255101
Add to Cart
Add to Cart
Type-H with Methanol vials, for identification of low concentration chemicals.
Kit includes 100 test sticks, and 100 1 mL methanol vials, compatible with LowDoseID enhanced Gemini.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Test Kit
Display No Display
For Use With (Equipment) Gemini Chemical Identification Analyzer