Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GemPure™ Cell and Molecular Grade Water, GeminiBio™
SDP

Catalog No. 11852120 Shop All Gemini Bio Products
Change view
Click to view available options
Quantity:
6 x 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-852-120 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For further manufacturing or research use only. Not for direct Therapeutic use.
Catalog No. 11-852-120 Supplier GeminiBio Supplier No. 9010041X6
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For further manufacturing or research use only. Not for direct Therapeutic use.

Manufactured in an ISO 13485 and 21 CFR 820 compliant facility and available in 1 L and custom sizes.

GemPure™ Cell and Molecular Grade Water is manufactured specifically for stringent research applications requiring high purity and low endotoxin levels. Sterile and free of proteases and nucleases, this grade supports both cell culture and molecular biology workflows while simplifying purchasing and conserving lab space.

  • Sterile and low endotoxin for sensitive cell culture applications
  • Protease-, DNase-, and RNase-free
  • Tested for sterility, endotoxin, TOC, and enzymatic contaminants

Suited for:

  • Cell culture workflows requiring low endotoxin water
  • Molecular biology applications sensitive to enzymatic contamination
  • Research teams seeking a single, versatile high-purity water solution

Chemical Identifiers

CAS 7732-18-5

Specifications

pH 5.0 to 7.0
Color Clear and Colorless
Conductivity <5.0 μS/cm
DNase DNase-free
Endotoxin <0.25 EU/mL
EU Ph Reference 169
Molecular Weight (g/mol) 18
ChEBI 15377
Percent Purity 0.99999
Grade Molecular Biology
Concentration or Composition (by Analyte or Components) H2O
Sterility Sterile
Filtered Through 0.2 μM Sterilization Grade Filter
Density 1 g/mL
Certificate of Analysis Yes
Quantity 6 x 1 L
Purification Method Filtration RO/DI
Shipping Condition Ambient protect from freezing
Chemical Name or Material Water, Water For Injection Quality
Show More Show Less
ShelfLife 24 Months
EINECSNumber 231-791-2
Recommended Storage 2°C to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.