Learn More
Perkin Elmer US LLC GemTip Cross-Flow II Nebulizer for Optima 2x00/4x00/5x00/7x00
Catalog No. 509043793
Encompass_Preferred
This nebulizer uses chemically resistant GemTips made of sapphire and ruby in a chemically resistant end cap (N0690676). It is included in the Quick-Change Torch Module with Scott Spray Chamber and GemTip Cross-Flow II Nebulizer for Optima 2x00/4x00/5x00/7x00 DV (N0770606) and the Scott/Cross-Flow II Sample Introduction Kit for Optima 2x00/4x00/5x00/7x00 DV (N0770731).
Catalog No. 50-904-3793 Supplier Perkin Elmer US LLC Supplier No. N0770546
This nebulizer uses chemically resistant GemTips made of sapphire and ruby in a chemically resistant end cap (N0690676). It is included in the Quick-Change Torch Module with Scott Spray Chamber and GemTip Cross-Flow II Nebulizer for Optima 2x00/4x00/5x00/7x00 DV (N0770606) and the Scott/Cross-Flow II Sample Introduction Kit for Optima 2x00/4x00/5x00/7x00 DV (N0770731).