Learn More
Perkin Elmer US LLC GemTip Cross-Flow II Nebulizer Brass Ferrule Back 1/4
Catalog No. 502113710
Encompass_Preferred
This Ferrule serves as a replacement part for the GemTip Cross-Flow II Nebulizer for Optima 8x00 and Avio 200/500 (N0780546) and the GemTip Cross-Flow II Nebulizer for Optima 2x00/4x00/5x00/7x00 (N0770546). The GemTip Cross-Flow II Nebulizer Ferrule Front 1/4 BRS (09903103) can be purchased separately.
Catalog No. 50-211-3710 Supplier Perkin Elmer US LLC Supplier No. 09903104
This Ferrule serves as a replacement part for the GemTip Cross-Flow II Nebulizer for Optima 8x00 and Avio 200/500 (N0780546) and the GemTip Cross-Flow II Nebulizer for Optima 2x00/4x00/5x00/7x00 (N0770546). The GemTip Cross-Flow II Nebulizer Ferrule Front 1/4 BRS (09903103) can be purchased separately.