Learn More
Perkin Elmer US LLC GemTip Cross-Flow II Nebulizer for Optima 8x00 and Avio 200/500
Catalog No. 502104126
Encompass_Preferred
This GemTip Cross-Flow II Nebulizer can routinely handle 50% (v/v) solutions of HCl, HNO3, H2SO4, H2PO4, 20% (v/v) HF and 30% (v/v) NaOH. Up to 20% NaCl can be aspirated by the nebulizer for 1 hour without clogging. It uses chemically resistant GemTips made of sapphire and ruby in a chemically resistant end cap.
Catalog No. 50-210-4126 Supplier Perkin Elmer US LLC Supplier No. N0780546
This GemTip Cross-Flow II Nebulizer can routinely handle 50% (v/v) solutions of HCl, HNO3, H2SO4, H2PO4, 20% (v/v) HF and 30% (v/v) NaOH. Up to 20% NaCl can be aspirated by the nebulizer for 1 hour without clogging. It uses chemically resistant GemTips made of sapphire and ruby in a chemically resistant end cap.