Learn More
MSC Gemtor Roofer's Fall Protection Kit, Universal, 300 lb, 1 ea
Catalog No. 5000795076
Encompass
This roofer's fall protection kit provides comprehensive safety for working at heights. It includes essential personal protective equipment designed for easy use and reliable performance in roofing applications.
  • Comprehensive fall protection kit for roofers.
  • Constructed from durable polyester material.
  • Kit includes: carrying bag, full body harness, lanyard, lifeline, roof anchor, and rope grab.
  • Features a 3 ft lanyard for secure attachment.
  • Equipped with a reusable roof anchor for multiple uses.
  • A 50 ft long, 5/8 in diameter rope lifeline is included.
Catalog No. 50-007-95076 Supplier MSC Supplier No. 81987919
This roofer's fall protection kit provides comprehensive safety for working at heights. It includes essential personal protective equipment designed for easy use and reliable performance in roofing applications.
  • Comprehensive fall protection kit for roofers.
  • Constructed from durable polyester material.
  • Kit includes: carrying bag, full body harness, lanyard, lifeline, roof anchor, and rope grab.
  • Features a 3 ft lanyard for secure attachment.
  • Equipped with a reusable roof anchor for multiple uses.
  • A 50 ft long, 5/8 in diameter rope lifeline is included.