Learn More
Analytical Sales & Services Gen II, 24 Position, Parallel Synthesis Reaction Block for 15mm OD (1 Dram) Vials, with Temperature Transfer Cover
Catalog No. 5000806976
Encompass
Gen II, 24 Position, Parallel Synthesis Reaction Block for 15mm OD (1 Dram) Vials, with Temperature Transfer Cover\n• For 15mm OD (1 Dram) Vials\n• 18mm Well Spacing\n• Includes Vial Rack, Temperature Transfer Cover and Screws (vials not included)
Catalog No. 50-008-06976 Supplier Analytical Sales & Services Supplier No. 104250
Gen II, 24 Position, Parallel Synthesis Reaction Block for 15mm OD (1 Dram) Vials, with Temperature Transfer Cover\n• For 15mm OD (1 Dram) Vials\n• 18mm Well Spacing\n• Includes Vial Rack, Temperature Transfer Cover and Screws (vials not included)