Learn More
Analytical Sales & Services Gen II HotSleeve+ Column Heater, Self-Calibrating, 30 cm Length, 5ºC to 130ºC Range, Unit
Supplier:  Analytical Sales & Services HS230L
Encompass
The Gen II HotSleeve+ is a innovative column heater designed for precise temperature control of analytical columns. It features self-calibration, a wide temperature range of 5ºC above ambient to 130ºC, and improved design for better fit. Compatible with MULTICON™ and MONOCON™ controllers, it excels in enhanced performance thanks to high-temperature VELCRO. Perfect for diverse scientific applications requiring consistent temperature, it differentiates itself with robust build and effective calibration.
Catalog No. NC2876279