Learn More
Analytical Sales & Services Gen II Powder Funnel Plate for 4x21mm Vials
Catalog No. 5000806111
Encompass
Gen II Powder Funnel Plate\nUse with Para-dox Gen II 96-position Reactors for 4x21mm Vials\n\n• Easily dispenses powder into your 4 x 21 micro vials\n• Extra wide openings for easy filling\n• Funneled holes guide powder down into vials\n• Circular grooves on underside of transfer plate “lock" vials into position to eliminate spillage and mess\n• Can be used with 96-Well Stackable Trays with 4x21mm Glass Inserts (96342)\n• Teflon Coating\n• Aluminum\n• Can also be used with Para-dox Gen I 96-position Reactors for 4x21mm Vials
Catalog No. 50-008-06111 Supplier Analytical Sales & Services Supplier No. 96T200
Gen II Powder Funnel Plate\nUse with Para-dox Gen II 96-position Reactors for 4x21mm Vials\n\n• Easily dispenses powder into your 4 x 21 micro vials\n• Extra wide openings for easy filling\n• Funneled holes guide powder down into vials\n• Circular grooves on underside of transfer plate “lock" vials into position to eliminate spillage and mess\n• Can be used with 96-Well Stackable Trays with 4x21mm Glass Inserts (96342)\n• Teflon Coating\n• Aluminum\n• Can also be used with Para-dox Gen I 96-position Reactors for 4x21mm Vials