Learn More
Thermco Products Inc General lab Non-Hg thermometer, Spirit, -30 deg./120 deg.F, 1 deg.F, 76mm imm,
Catalog No. NC0609473
Encompass_Preferred
Spirit filled double scale thermometer, white back tubing, with toughened bulbs. Suspension ring for hanging, vertical numbering. Individual serial numbered. Packed in non-roll square tubing. Permanent markings.
Catalog No. NC0609473 Supplier Thermco Products Inc Supplier No. B120FW3S
Spirit filled double scale thermometer, white back tubing, with toughened bulbs. Suspension ring for hanging, vertical numbering. Individual serial numbered. Packed in non-roll square tubing. Permanent markings.