Learn More
Waters Corp Waters Anion-Exchange Column, 2.5 μm, 4.6 mm, 100 mm, Ion-Exchange
Catalog No. 50785078 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Gen-Pak FAX columns offer high resolution for anion-exchange HPLC of nucleic acids. These columns contain a weak anion exchanger based on DEAE functionalized non-porous resin with 2.5 μm spherical particles, suitable for analytical and micro-preparative applications. Measuring 4.6 mm in inner diameter and 100 mm in length, they are constructed from steel and designed for high-resolution analysis and purification of various nucleic acid species.
  • High resolution for anion-exchange HPLC of nucleic acids.
  • Weak anion exchanger based on DEAE functionalized non-porous resin.
  • 2.5 μm spherical particles for analytical and micro-preparative use.
  • Dimensions: 4.6 mm inner diameter and 100 mm length.
  • Constructed from steel.
  • Designed for high-resolution analysis and purification of nucleic acid species.
  • pH operating range of 2 to 8.
  • Maximum pressure of 6000 psi (415 bar).
Catalog No. 50-785-078 Supplier Waters Corp Supplier No. WAT015490
Gen-Pak FAX columns offer high resolution for anion-exchange HPLC of nucleic acids. These columns contain a weak anion exchanger based on DEAE functionalized non-porous resin with 2.5 μm spherical particles, suitable for analytical and micro-preparative applications. Measuring 4.6 mm in inner diameter and 100 mm in length, they are constructed from steel and designed for high-resolution analysis and purification of various nucleic acid species.
  • High resolution for anion-exchange HPLC of nucleic acids.
  • Weak anion exchanger based on DEAE functionalized non-porous resin.
  • 2.5 μm spherical particles for analytical and micro-preparative use.
  • Dimensions: 4.6 mm inner diameter and 100 mm length.
  • Constructed from steel.
  • Designed for high-resolution analysis and purification of nucleic acid species.
  • pH operating range of 2 to 8.
  • Maximum pressure of 6000 psi (415 bar).