Learn More
MSC Rochester Midland Corporation Cleaner and Degreaser, Liquid, Concentrated, 5 gal
Catalog No. 502816630
Encompass
Rochester Midland Corporation CLEANAIRE 1200 is a concentrated, low-odor, liquid cleaner and degreaser. This water-based solution is effective for removing oil, grease, and grease gum from surfaces such as parts, machinery, floors, walls, and vehicles.
  • Unique surfactant system floats soils away
  • High foaming reduces labor
  • Soft metal safe due to inhibition
  • Biodegradable and free of butyl, phosphate, and caustic
  • Economical with high dilution ratios (1:10 to 1:128)
  • Free rinsing, preventing filming
  • Safe on paint and plastics
  • Low VOC content (0 g/L)
  • High pH of 12.5 +/- 0.5
Catalog No. 50-281-6630 Supplier MSC Supplier No. 32788606
Rochester Midland Corporation CLEANAIRE 1200 is a concentrated, low-odor, liquid cleaner and degreaser. This water-based solution is effective for removing oil, grease, and grease gum from surfaces such as parts, machinery, floors, walls, and vehicles.
  • Unique surfactant system floats soils away
  • High foaming reduces labor
  • Soft metal safe due to inhibition
  • Biodegradable and free of butyl, phosphate, and caustic
  • Economical with high dilution ratios (1:10 to 1:128)
  • Free rinsing, preventing filming
  • Safe on paint and plastics
  • Low VOC content (0 g/L)
  • High pH of 12.5 +/- 0.5