Learn More
Research Products International Corp General Purpose Sealing Film, Non-Sterile, 100pk
SDP
Catalog No. 502253051
Encompass
General Purpose polypropylene film offers superior barrier properties to minimize evaporation and prevent well-to-well contamination. Recommended for general laboratory use, short-term storage, incubation and ELIZA. Provides a very secure seal to minimize evaporation and is slightly harder to remove than SealPlate films. Manufactured with polypropylene that reduces aerosols upon removal and features good chemical resistance. Specially formulated adhesive minimizes plate residue. Certified DNase and RNase free and -40·C to +120°C temperature range.
Certified DNase and RNase free
Temperature range -40°C to +120°C
Reduces aerosols upon removal
This item is not returnable. View return policy
Catalog No. 50-225-3051 Supplier Research Products International Corp Supplier No. 202542
General Purpose polypropylene film offers superior barrier properties to minimize evaporation and prevent well-to-well contamination. Recommended for general laboratory use, short-term storage, incubation and ELIZA. Provides a very secure seal to minimize evaporation and is slightly harder to remove than SealPlate films. Manufactured with polypropylene that reduces aerosols upon removal and features good chemical resistance. Specially formulated adhesive minimizes plate residue. Certified DNase and RNase free and -40·C to +120°C temperature range.
Certified DNase and RNase free
Temperature range -40°C to +120°C
Reduces aerosols upon removal