Learn More
MSC Guardair General Purpose Air Hose Assembly, 3/4 in ID, 25 ft, 200 psi
Catalog No. 502835612
Encompass
This tough, abrasion- and ozone-resistant, heavy-duty rubber air hose is designed to supply compressed air to safety air guns and vacuums. It includes Universal fittings on each end and is ideal for supplying compressed air to vacuums in various applications.
  • Minimum pressure rating: 200 psi
  • Pre-installed universal couplings
  • Swivel male NPT fittings on both ends
  • Fittings: 3/4 in MNPT swivel x 3/4 in MNPT swivel
Catalog No. 50-283-5612 Supplier MSC Supplier No. 74768623
This tough, abrasion- and ozone-resistant, heavy-duty rubber air hose is designed to supply compressed air to safety air guns and vacuums. It includes Universal fittings on each end and is ideal for supplying compressed air to vacuums in various applications.
  • Minimum pressure rating: 200 psi
  • Pre-installed universal couplings
  • Swivel male NPT fittings on both ends
  • Fittings: 3/4 in MNPT swivel x 3/4 in MNPT swivel