Learn More
MSC GEN PURPOSE VINYL TAPE
Catalog No. 5000796652
Encompass
3M General Purpose Vinyl Tape 764 is a cost-effective vinyl tape with a rubber adhesive, ideal for safety marking and color-coding. Its conformable backing allows it to adhere to irregular surfaces without lifting, and it maintains vivid color even when exposed to abrasion and solvents.
  • Cost-effective for color coding and safety marking
  • Maintains vivid color against abrasion and solvents
  • Conformable backing for curved and irregular surfaces
  • Rubber adhesive provides instant adhesion
  • Suitable for light-duty floor marking and general bundling
Catalog No. 50-007-96652 Supplier MSC Supplier No. 04582656
3M General Purpose Vinyl Tape 764 is a cost-effective vinyl tape with a rubber adhesive, ideal for safety marking and color-coding. Its conformable backing allows it to adhere to irregular surfaces without lifting, and it maintains vivid color even when exposed to abrasion and solvents.
  • Cost-effective for color coding and safety marking
  • Maintains vivid color against abrasion and solvents
  • Conformable backing for curved and irregular surfaces
  • Rubber adhesive provides instant adhesion
  • Suitable for light-duty floor marking and general bundling