Learn More
MSC GEN PURPOSE VINYL TAPE
Catalog No. 5000788500
Encompass
Floor & Aisle Marking Tape is a light-duty vinyl tape designed for marking applications in light traffic areas. It features a non anti-slip surface and maintains vivid color even when exposed to abrasion and solvents, making it suitable for a variety of environments.
  • Conformable backing for use on curved and irregular surfaces
  • Rubber adhesive provides good instant adhesion on many surfaces
  • Can be applied by hand or with a floor applicator
  • Complies with ASTM specifications
  • Maintains vivid color even when exposed to abrasion and solvents
  • Non anti-slip surface
  • Light-duty traffic type
Catalog No. 50-007-88500 Supplier MSC Supplier No. 04582631
Floor & Aisle Marking Tape is a light-duty vinyl tape designed for marking applications in light traffic areas. It features a non anti-slip surface and maintains vivid color even when exposed to abrasion and solvents, making it suitable for a variety of environments.
  • Conformable backing for use on curved and irregular surfaces
  • Rubber adhesive provides good instant adhesion on many surfaces
  • Can be applied by hand or with a floor applicator
  • Complies with ASTM specifications
  • Maintains vivid color even when exposed to abrasion and solvents
  • Non anti-slip surface
  • Light-duty traffic type