Learn More
Genscript Corporation GenBox 10 10 Buffer Dam
Catalog No. 5000057770
Encompass
10 10 buffer dam for GenBox Mini Electrophoresis Tank One-piece buffer dam is used when running only one gel
Catalog No. 50-000-57770 Supplier Genscript Corporation Supplier No. L00775
10 10 buffer dam for GenBox Mini Electrophoresis Tank One-piece buffer dam is used when running only one gel