Learn More
Genscript Corporation GenBox Mini Blot module
Catalog No. 5000058404
Encompass
The GenBox Mini Blot Module is compatible with GenBox Mini Electrophoresis tank which is interchangeable with the electrophoresis module The GenBox Mini Trans- Blot module accommodates two gel holder cassettes which is useful for blotting 2 mini PAGE gels at the same time
Catalog No. 50-000-58404 Supplier Genscript Corporation Supplier No. L00781
The GenBox Mini Blot Module is compatible with GenBox Mini Electrophoresis tank which is interchangeable with the electrophoresis module The GenBox Mini Trans- Blot module accommodates two gel holder cassettes which is useful for blotting 2 mini PAGE gels at the same time