Learn More
Genscript Corporation GenBox Mini Blot System
Catalog No. 5000058413
Encompass
The GenBox Mini Blot System is compatible with both the GenBox Mini electrophoresis module as well as the blot modules for tank blotting The system holds both GenScript s 10 8 and 10 10 precast gels interchangeably
Catalog No. 50-000-58413 Supplier Genscript Corporation Supplier No. L00782
The GenBox Mini Blot System is compatible with both the GenBox Mini electrophoresis module as well as the blot modules for tank blotting The system holds both GenScript s 10 8 and 10 10 precast gels interchangeably