Learn More
Genscript Corporation GenBox Mini Electrophoresis ta
Catalog No. 5000059125
Encompass
The GenBox Mini Electrophoresis Tank is compatible with GenScript s 10 8 precast gels and can run one or two mini gels The mini tank also works with GenBox mini blot modules for tank blotting
Catalog No. 50-000-59125 Supplier Genscript Corporation Supplier No. L00780
The GenBox Mini Electrophoresis Tank is compatible with GenScript s 10 8 precast gels and can run one or two mini gels The mini tank also works with GenBox mini blot modules for tank blotting