Learn More
Genscript Corporation GenBox Wedge
Catalog No. 5000057416
Encompass
Wedge is used for holding precast gel in place
Catalog No. 50-000-57416 Supplier Genscript Corporation Supplier No. L00784
Wedge is used for holding precast gel in place