Learn More
Genscript Corporation GenBuilder Plus Cloning Kit
Catalog No. 5000057423
Encompass
The GenBuilder Plus Cloning Kit enables seamless and one-step cloning of multiple DNA fragments into a linearized vector The proprietary enzyme mix fuses together DNA fragments efficiently and precisely by recognizing 15-40 bp overlaps at the end of each fragment The system allows cloning of DNA fragments into any linearized vector requires no pre-existing recombination sites or helper sequences and eliminates the need for a complicated restriction and ligation process The GenBuilder Plus Cloning kit can simultaneously clone 12 DNA fragments into a linearized vector The kit can also be used to assemble single stranded DNA (ssDNA) oligos with double stranded DNA (dsDNA) fragments and using ssDNA as a linker to fuse together two adjacent dsDNA fragments
Catalog No. 50-000-57423 Supplier Genscript Corporation Supplier No. L0074410
The GenBuilder Plus Cloning Kit enables seamless and one-step cloning of multiple DNA fragments into a linearized vector The proprietary enzyme mix fuses together DNA fragments efficiently and precisely by recognizing 15-40 bp overlaps at the end of each fragment The system allows cloning of DNA fragments into any linearized vector requires no pre-existing recombination sites or helper sequences and eliminates the need for a complicated restriction and ligation process The GenBuilder Plus Cloning kit can simultaneously clone 12 DNA fragments into a linearized vector The kit can also be used to assemble single stranded DNA (ssDNA) oligos with double stranded DNA (dsDNA) fragments and using ssDNA as a linker to fuse together two adjacent dsDNA fragments