Learn More
Epoch Life Science Inc GenCatch™ Advanced PCR Cleanup Kit (250 preps) Uniquely formulated buffers, reinforced columns with collection tubes, along with protocols.
SDP
Supplier:  Epoch Life Science Inc 2360250
Encompass_Preferred
PCR purification kit. Silica membrane spin column based. Sample volume: 850 ul. Column binding capacity: 40 ug DNA. Minimum elution volume: 20ul. 18 month shelf life.
Catalog No. NC0379253