Learn More
Genscript Corporation Antibody Enzyme Other Products Molecular Biology 100 UG 1/EA
Supplier:  Genscript Corporation Z03502100
Encompass
Recombinant Acidaminococcus sp. BV3L6 Cas12a (cpf1) nuclease is expressed in E. coli and purified. The nuclease contains nuclear localization sequence (NLS) at the C-terminus and 6× His-tag at the C-terminus.
Catalog No. 50-252-9623