Learn More
Genscript Corporation GenCRISPR™ Cas13a (C2c2) Nuclease
Supplier:  Genscript Corporation Z03486500
Encompass
GenCRISPR Cas13a (C2c2) Nuclease is an RNA-guided, RNA-targeting CRISPR enzyme, commonly referred to as C2c2. This enzyme is a member of Type VI CRISPR family containing a single protein effector with two HEPN domains. The Cas13a exhibits two RNase activities. The first activity, a cis- sequence-specific RNA guided cleavage, activates the secondary RNase activity of the enzyme, a non-specific trans- ribonuclease activity. This Cas13a protein can be applied to detect single RNA molecules with proven high specificity.
Catalog No. NC2011092