Learn More
Genscript Corporation Antibody Enzyme Other Products Molecular Biology 500 UG 1/EA
Supplier:  Genscript Corporation Z03702500
Encompass
GenCRISPR Cas9 v1.2 is a tag free nuclease produced by expression in an E. coli strain carrying a plasmid encoding the Cas9 gene from Streptococcus pyogenes with a biparticle nucleus localization signal (BPNLS) at N-terminal and a nucleoplasmin nucleus localization signal (nucleoplasmin NLS) at C-terminal. It has been reported that BPNLS and nucleoplasmin NLS are able to improve the gene editing efficiency.
Catalog No. 50-252-9576