Learn More
Genscript Corporation Antibody Enzyme Other Products Molecular Biology 1 MG 1/EA
Supplier:  Genscript Corporation Z037421
Encompass
GenCRISPR LbuCas13a Nuclease is an RNA-guided RNA-targeting CRISPR enzyme commonly referred to as LbuC2c2. This Cas13a protein can be applied to detect single RNA molecules with proven high specificity. Recently Cas13a has been leveraged as a diagnostic nucleic acid detection tool.
Catalog No. 50-252-9567