Learn More
Genscript Corporation Antibody Enzyme Other Products Molecular Biology 100 UG 1/EA
Supplier:  Genscript Corporation Z03467100
Encompass
GenCrispr NLS-Cas9-EGFP is a fusion protein developed by GenScript. It contains a nuclear localization sequence (NLS) on its N terminal and EGFP on the C terminal.
Catalog No. 50-252-9626