Learn More
Genscript Corporation Antibody Enzyme Other Products Molecular Biology 50 UG 1/EA
Supplier:  Genscript Corporation Z0339350
Encompass
GenCrispr NLS-Cas9-EGFP is a fusion protein developed by GenScript. It contains a nuclear localization signal (NLS) on its N terminal end and an EGFP and a 6x(His) sequence on the C terminal end.
Catalog No. 50-252-9638