Learn More
Genscript Corporation GenCrispr NLS-Cas9-EGFP Nuclease
Supplier:  Genscript Corporation Z03393100
Encompass
GenCrispr NLS-Cas9-EGFP is a fusion protein developed by GenScript. It contains a nuclear localization signal (NLS) on its N terminal end, and an EGFP and a 6x(His) sequence on the C terminal end. Cas9 protein forms a highly stable ribonucleoprotein (RNP) complex with the guide RNA (gRNA) component of the CRISPR/Cas9 system. When Cas9 is expressed with an NLS sequence, the Cas9 RNP complex can localize to the nucleus immediately upon entering the cell. There is no requirement for in vivo transcription or translation, which improves the efficiency of this method over plasmid-based systems. The EGFP tag can be used as a reporter for tracking or sorting transfected cells, which enables enrichment of cell populations for desired genome edits via fluorescence activated cell sorting (FACS). It significantly reduces the labor and cost associated with single cell cloning and genotyping in genome editing applications.
Catalog No. NC1438302