Learn More
Genscript Corporation Antibody Enzyme Other Products Molecular Biology 1 MG 1/EA
Supplier:  Genscript Corporation Z036991
Encompass
GenCRISPR SaCas9 2NLS Nuclease is a tag free nuclease produced by expression in an E. coli strain carrying a plasmid encoding the Cas9 gene from Staphylococcus aureus with a nuclear localization signal at both N-terminal and C-terminal.The small size of the nuclease facilitates enhanced in vivo delivery for genome editing in various organisms.
Catalog No. 50-252-9579