Learn More
Genscript Corporation GenCRISPR™ SaCas9 2NLS Nuclease
Supplier:  Genscript Corporation Z036990.1
Encompass
GenCRISPR™ SaCas9 2NLS Nuclease is a tag free nuclease produced by expression in an E. coli strain carrying a plasmid encoding the Cas9 gene from Staphylococcus aureus with a nuclear localization signal at both N-terminal and C-terminal.The small size of the nuclease facilitates enhanced in vivo delivery for genome editing in various organisms.
Catalog No. NC2050153