Learn More
Genscript Corporation GenCrispr T7 Endonuclease I
Supplier:  Genscript Corporation Z03396250
Encompass
T7 Endonuclease I (T7E1) recognizes and cleaves non-perfectly matched DNA cruciform DNA structures Holliday structures or junctions hetero duplex DNA and more slowly nicked double-stranded DNA The cleavage site is at the first second or third phosphodiester bond that is 5 to the mismatch The protein is the product of T7 gene 3 GenCrispr T7 Endonuclease I is a fusion protein produced from i E coli /i
Catalog No. 50-000-58526