Learn More
Genscript Corporation GenCRISPR™ Ultra eSpCas9-N-NLS Research, tag-free
Supplier:  Genscript Corporation Z036920.5
Encompass
GenCRISPR™ Ultra eSpCas9-N-NLS Research, tag-free is a mutant form of Cas9 nuclease and is produced by expression in an E. coli strain carrying a plasmid encoding the eSpCas9 gene from Streptococcus pyogenes with an N-terminal nuclear localization signal (N-NLS). GenCRISPR™ Ultra eSpCas9-N-NLS Research, tag-free delivers higher fidelity and less off-target activity than wild-type SpCas9 nuclease.
Catalog No. NC2050152