Learn More
GENTUERI INC GenDry™ Pouches for CollectEjec™ Pack of 100
SDP
Catalog No. NC2146456
Encompass_Preferred
The GenDry™ Pouch is a sealed foiled pouch with a zip closure for preserving samples after collection to transport to the laboratory for processing. GenDry™ CE: Sealed pouch includes a 2g desiccant and is designed to accept a CollectEject™ Swab.
This item is not returnable. View return policy
Catalog No. NC2146456 Supplier GENTUERI INC Supplier No. GD400NS
The GenDry™ Pouch is a sealed foiled pouch with a zip closure for preserving samples after collection to transport to the laboratory for processing. GenDry™ CE: Sealed pouch includes a 2g desiccant and is designed to accept a CollectEject™ Swab.