Learn More
Agilent Technologies GENE MORPH II EZ CLONE KIT
Catalog No. NC9297817
Encompass
GeneMorph II EZClone Domain Mutagenesis Kit; 3-step cloning method which eliminates tedious sub-cloning and the need for specific restriction sites; Yields of targets up to 3.5 kb; Control mutation frequency from 1 to 16 mutations per kb
Catalog No. NC9297817 Supplier Agilent Technologies Supplier No. 200552
GeneMorph II EZClone Domain Mutagenesis Kit; 3-step cloning method which eliminates tedious sub-cloning and the need for specific restriction sites; Yields of targets up to 3.5 kb; Control mutation frequency from 1 to 16 mutations per kb