Learn More
Bio Rad Laboratories GENE PULSER CUVETTE (.1CM)
Catalog No. NC9243229
Encompass
Gene Pulser Cuvette Pack, 50pk, Standard, Sterile, Individually wrapped, 0.1cm electrode gap
Catalog No. NC9243229 Supplier Bio Rad Laboratories Supplier No. 1652089
Gene Pulser Cuvette Pack, 50pk, Standard, Sterile, Individually wrapped, 0.1cm electrode gap