Learn More
Bio Rad Laboratories GENE PULSER CUVETTE (.4CM)
Catalog No. NC9327166
Encompass
Gene Pulser Cuvette Pack, 50pk, Standard, Sterile, Individually wrapped, 0.4cm electrode gap
Catalog No. NC9327166 Supplier Bio Rad Laboratories Supplier No. 1652088
Gene Pulser Cuvette Pack, 50pk, Standard, Sterile, Individually wrapped, 0.4cm electrode gap