Learn More
Bio Rad Laboratories GENE PULSER CUVETTES
Catalog No. NC9105095
Encompass
Gene Pulser/MicroPulser Cuvettes, 0.2cm gap, 5 (mini pack)
Catalog No. NC9105095 Supplier Bio Rad Laboratories Supplier No. 1652082
Gene Pulser/MicroPulser Cuvettes, 0.2cm gap, 5 (mini pack)