Learn More
Bio Rad Laboratories GENE PULSER/MICROPULSER CUVETT
Catalog No. 509011774
Encompass
cuvet; Bio-Rad; Gene Pulser/MicroPulser; 0.1cm gap; Pack of 500
Catalog No. 50-901-1774 Supplier Bio Rad Laboratories Supplier No. 1652093
cuvet; Bio-Rad; Gene Pulser/MicroPulser; 0.1cm gap; Pack of 500