Learn More
Bio Rad Laboratories GENE PULSER/MICROPULSER CUVETT
Catalog No. 509011775
Encompass
Cuvet; Bio-Rad; Gene Pulser/MicroPulser; 0.2cm gap; Pack of 500
Catalog No. 50-901-1775 Supplier Bio Rad Laboratories Supplier No. 1652092
Cuvet; Bio-Rad; Gene Pulser/MicroPulser; 0.2cm gap; Pack of 500