Learn More
Bio Rad Laboratories GENE PULSER XCELL PC MODULE
Catalog No. 509010698
Encompass
PC Module; Bio-Rad; Gene Pulser Xcell; Modular electroporation system; Exponential decay delivery; Recommended for bacteria, fungi, other applications; High-voltage pulses applied; 100/240 V, 50/60 Hz; 31 x 28 x 5cm; wt.:1.9 kg
Catalog No. 50-901-0698 Supplier Bio Rad Laboratories Supplier No. 1652668
PC Module; Bio-Rad; Gene Pulser Xcell; Modular electroporation system; Exponential decay delivery; Recommended for bacteria, fungi, other applications; High-voltage pulses applied; 100/240 V, 50/60 Hz; 31 x 28 x 5cm; wt.:1.9 kg