Learn More
Bio Rad Laboratories GENE PULSER XCELL SHOCKPOD CUV
Catalog No. 509010699
Encompass
ShockPod; Bio-Rad Laboratories; Cuvette chamber; For Gene Pulser Xcell; 100/240 V, 50/60 Hz; Includes integral leads for connection to Gene Pulser Xcell, Gene Pulser II, or MicroPulser electroporators
Catalog No. 50-901-0699 Supplier Bio Rad Laboratories Supplier No. 1652669
ShockPod; Bio-Rad Laboratories; Cuvette chamber; For Gene Pulser Xcell; 100/240 V, 50/60 Hz; Includes integral leads for connection to Gene Pulser Xcell, Gene Pulser II, or MicroPulser electroporators