Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GeneArt™ Cryopreservation Kit for Algae

Catalog No. A24228
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24228 100 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A24228 Supplier Invitrogen™ Supplier No. A24228
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to preserve algal strains and clones for storage at -80°C for years, thus eliminating liquid nitrogen storage and continuous cultures as a way to maintain your clones

The GeneArt™ Cryopreservation Kit for Algae is used to preserve algal strains and clones for storage at -80°C for years, thus eliminating liquid nitrogen storage and continuous cultures as a way to maintain your clones. Cryopreservation is the optimal choice for preservation and long-term storage of algae, since it minimizes genetic drift, facilitates strain and clone exchange between labs, and helps reduce maintenance labor and costs. Simply grow your cells in the presence of Cryopreservation Reagent A for 2–5 days, harvest, resuspend in Cryopreservation Reagent B, and freeze a small aliquot in one of two recommended freezing containers* at -80°C. The GeneArt™ Cryopreservation Kit for Algae has been used to preserve Chlamydomonas and Chlorella strains with 100% resuscitation after thawing.
  • Preserve Chlamydomonas and Chlorella strains and clones for -80°C storage
  • Eliminate liquid nitrogen storage
  • Minimize contamination and genetic drift that typically occur in continuous cultures
  • Ship algal clones on dry ice for exchange between labs

To date, most available methods for algae cell preservation have required liquid nitrogen storage, but this method is inconvenient and expensive. Alternatively, researchers have relied on continuous cultures to maintain algal clones, but this often leads to contamination and genetic drift and is not amenable to shipping. The GeneArt™ Cryopreservation Kit for Algae provides a way to freeze algae cells at -80°C for long-term storage or shipment on dry ice with minimized loss in viability.

*We have used the following freezing containers with this product:
  • Thermo Scientific™ Mr. Frosty™ Freezing Container
  • Biocision™ CoolCell™ FTS30 Cell Freezing Container

For Research Use Only. Not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Algae Cryopreservation Kit
Product Line GeneArt
Quantity 100 preps
Species Chlamydomonas
Content And Storage Contains:
• 1 bottle Cryopreservation Reagent A, 5 mL
• 1 bottle Cryopreservation Reagent B, 25 mL
Expression System Algae
What freezing container should I use along with the GeneArt Cryopreservation Kit?

We have used the following freezing containers with this product:

Thermo Scientific Mr. Frosty Freezing Container
Biocision CoolCell FTS30 Cell Freezing Container

What is the cell viability when using the GeneArt Cyropreservation Kit?

We have tested the kit with Chlamydomonas and Chlorella strains, showing a 5% and 100% cell viability, respectively.

How does the GeneArt Cryopreservation Kit work?

Simply grow your cells in the presence of Cryopreservatin Reagent A for 2-5 days, harvest the cells, and resuspend them in Cryopreservation Reagent B. Follow by freezing the cells at -80 degrees celsius.

Do you offer a kit for the preservation of algal strains and clones?

Yes, we offer our GeneArt Cryopreservation Kit for Algae (Cat. No. A24228).

What type of algae can the MAX Efficiency Transformation Reagent for Algae be used for?

The MAX Efficiency Transformation Reagent for Algae was developed for transforming the pChlamy_4 vector into the Chlamydomonas reinhardtii 137c cells by electroporation.

What type of algae can the GeneArt Cryopreservation Kit for Algae be used for?

The GeneArt Cryopreservation Kit for Algae was developed for preserving the Chlamydomonas reinhardtii 137c cells. We have reports from some customers who were successfully able to preserve other algae cells.

What does the GeneArt Chlamydomonas Protein Expression Vector include?

It includes the pChlamy_4 vector. This vector has been tested for protein expression in Chlamydomonas reinhardtii 137c . You can get the Chlamydomonas reinhardtii 137c cells from the Chlamydomonas Resource Center (http://www.chlamycollection.org/).


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.