Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GeneArt™ High-Order Genetic Assembly Systems, with yeast growth media

Catalog No. A13286
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13286 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A13286 Supplier Invitrogen™ Supplier No. A13286
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A highly efficient kit for the simultaneous and seamless assembly of up to 10 DNA fragments, totaling up to 110 Kbp in length, into any vector.

The GeneArt™ High-Order Genetic Assembly System is a highly efficient kit for the simultaneous and seamless assembly of up to 10 DNA fragments, totaling up to 110 Kbp in length, into any vector. The system relies on yeast’s ability to take up and recombine DNA fragments with high efficiency. This greatly reduces the in vitro handling of DNA and eliminates the need for enzymatic treatments, such as restriction and ligation, while allowing for precise fusions of DNA sequences. The kit contains materials for the transformation and purification from yeast (available with or without yeast selective media), and competent E. coli for plasmid amplification of correct clones.
  • Easy and Powerful — Clone up to 10 DNA fragments, with the sequence of your choice, simultaneously in a single vector (up to 110 Kbp); no restriction digestion, ligation or recombination sites required
  • Precision and Efficiency —Designed to let you clone what you want, where you want, in the orientation you want, and achieve up to 90% correct clones with no extra sequences left behind
  • Flexibility — Use our linear vector, a vector of your choice, or clone pre-existing DNA fragments that have no end-homology without further modifications
  • Free Tools — Design DNA oligos and more with our free web-based interface that walks you through your project step-by-step
  • Diverse Applications — Streamline many synthetic biology and molecular biology techniques through the rapid combination, addition, deletion, or exchange of DNA segments

For the cloning of 1 to 4 DNA fragments of limited size, and if you prefer an in vitro approach, consider using the GeneArt™ Seamless Cloning and Assembly Kit (cat # A13288).

Easily Create New Specific Constructs from Diverse DNA Fragments
The GeneArt™ High-Order Genetic Assembly System takes advantage of transformation-associated recombination (TAR) in the yeast Saccharomyces cerevisae to join pre-existing DNA fragments, or chemically synthesized oligonucleotides, into a single recombinant molecule. DNA fragments and linearized vector are joined based on shared end-terminal homology. If no such end homology exists between pieces, they can be “stitched" together with recombination linkers, synthetic DNA oligonucleotides that provide end-terminal homology between two unrelated DNA fragments. The process is very efficient and seamless, leaving no extra sequences after the assembly. Even though it has been shown to work for up to 0.5 Mb and 50 DNA fragments, this product has been optimized for up to 110 Kb and 10 DNA fragments in a vector.

Simple Clone Verification
In order to minimize the work that is done in yeast, recombinant yeast clones are subjected to a simplified 10-minute DNA extraction protocol. The extraction step yields assembled molecule in enough quantity to do colony PCR verification of the junctions, as well as direct transformation of E.coli cells for downstream analysis. The proprietary lysis buffer and glass beads needed for the extraction are included in the kit. For selection of recombinant yeast clones we offer a selective yeast media kit, CSM Media for Mav203 Yeast Cells (cat# A13292).

Considerations for Choosing a Cloning Vector
The GeneArt™ High-Order Genetic Assembly System requires shuttle vectors that have high capacity and are compatible with yeast and E.coli (i.e. BAC-YAC shuttle vectors). There is no cloning vector included in this product, but we offer a ready-to-use linear cloning vector separately called the GeneArt™ pYES1L Vector with Sapphire Technology™ (cat# A13287). You can also use your own vector, but it must be compatible with the High-Order Genetic Assembly System. This compatibility can be easily accomplished with our GeneArt™ High-Order Vector Conversion Cassette (cat#A13291).

Cloning Efficiency
In GeneArt™ High-Order Assembly the main factors affecting cloning efficiency are the size of the DNA elements, the number of those without end-homology, the total size of the final molecule, and the quality and specificity of the fragment. The terminal end of the PCR fragments (A-overhangs or blunt), does not affect the cloning efficiency.

Typical cloning efficiencies for different numbers of fragments with end-homology assembled into the GeneArt™ pYES1L Vector with Sapphire Technology™ are the following:
  • >90% for 5 DNA fragments of 10 Kb each
  • >90% for 10 DNA fragments of 5 Kb each
  • >50% for 10 DNA fragments of 10 Kb each

Common cloning efficiencies for pre-existing fragments, without end-homology, assembled into the GeneArt™ pYES1L Vector with Sapphire Technology™ using 'stitching’ DNA oligonucleotides are:
  • >90% for 1 fragment of 10 Kb
  • >75% for 2 DNA fragments of 10 Kb each

Design Your Cloning in silico
A key step in GeneArt™ High-Order Genetic Assembly is the correct design of fragments and oligos with the appropriate homology and spacing to ensure successful assembly of your clone. To simplify and speed the design process we provide a free online design tool to help you design your experiment in silico. The tool checks for compatibility of the experimental design with the product specifications, designs DNA oligos for either PCR amplification or for stitching of the different elements to clone, and presents the user with a graphical representation of the vector as well as a downloadable annotated sequence in GenBank format that is compatible with Vector NTI™ software.

Applications
The GeneArt™ High-Order Genetic Assembly System is designed to empower cloning and DNA assembly experiments in a wide range of molecular biology and synthetic biology applications, among others. The product allows for the creation of modular expression vectors, with interchangeable parts, and can be used to perform a variety of tasks that would otherwise involve multiple steps. Use the kit to simply: construct fusion proteins, delete, replace, or add DNA elements such as restriction sites, clone large pre-existing DNA fragments without end-homology, and many other techniques that require manipulation of genetic sequences.

For Research Use Only. Not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY

Specifications

Bacterial or Yeast Strain MaV203, TOP10
Cloning Method Seamless Cloning
Content And Storage The system has enough reagents for 10 assembly reactions and a control as well as the reagents to make 2 liters of yeast growth media. The components are: One Shot™ MaV203 yeast competent cells, One Shot™ TOP10 Electrocompetent E.coli cells, PEG⁄Lithium Acetate solution, S.O.C. medium, control cloning vector and insert for DNA assembly, supercoiled vector controls for transformation, lysis buffer, glass beads and two CSM agar media pouches and 2 bottles of sterile liquid 20% glucose to make 2 liters of CSM agar media.

A cloning vector is not included in this kit.

For your convenience, this product is also available without the yeast agar media under the catalog number A13285.

Store the -80°C module at -80°C and the rest of the reagents at room temperature (CSM-Trp media, 20% glucose, lyses buffer, glass beads and S.O.C. medium).
Description GeneArt High-Order Genetic Assembly System, with yeast growth media
Format Kit
Sample Type DNA
For Use With (Application) Cloning
Number of Fragments Up to 10 Fragments
Product Line GeneArt
Product Type Genetic Assembly System Kit
Quantity 10 Reactions
Size 110 kb total (vector plus all inserts)
Workflow Step DNA Assembly
Show More Show Less
With the GeneArt High-Order Genetic Assembly System, I'm getting no positive colonies detected by yeast colony PCR. Can you please offer some troubleshooting tips?

We would recommend trying to re-streak the colony on a fresh plate and repeat colony PCR. Do not break open the yeast cells with the beads supplied with the kit; the beads are for transformation into E. coli. Additionally, use less than 0.5 µL of diluted yeast lysate in a 50 µL PCR reaction.

With the GeneArt High-Order Genetic Assembly System, I see small or no yeast colonies after transformation. Can you please offer some suggestions?

Ensure that yeast transformations are incubated at 30 degrees C for 3 days for proper colony formation.

With the GeneArt High-Order Genetic Assembly System, I did not get any yeast colonies after transformation and the transformation control did not work. Can you please offer some suggestions?

Please review the following suggestions:
– Perform transformation exactly as described in protocol.
– Do not freeze-thaw or vortex MaV203 yeast competent cells.
– Use CSM-Trp agar plates for the transformation.
– For best results, use fresh DMSO from an unopened bottle. You may use DMSO stored at -20 degrees C.

I want to use my own E. coli vector for my assembly. Can I do this? How?

Yes, you should be able to adapt your E. coli vector into a yeast-compatible cloning vector using the GeneArt High-Order Vector Conversion Cassette (Cat. No. A13291) for use with the GeneArt High-Order Genetic Assembly System with the following provisions:
– Start by using the DNA Oligo Designer web tool, and verify that your vector and the GeneArt High-Order Vector Conversion Cassette do not share internal homology to prevent potential re-arrangements when using your adapted vector with the GeneArt High-Order Genetic Assembly System.
– Use a vector with a single- or low-copy-number origin for a final construct of >15 kb, if the final plasmid construct will be transferred into E. coli. Usually, low-copy-number E. coli vectors have significantly higher capacity than high-copy number vectors.
– Avoid chloramphenicol selection markers on the custom vector since this is the marker on the cassette.
– After ligation (1:10 vector: insert ratio recommended), transform competent E. coli cells with the ligation mixture and plate on double selection LB plates (chloramphenicol plus the antibiotic marker on your custom vector backbone).To linearize your yeast-adapted cloning vector for multi-fragment assembly, a double-digestion is required to avoid background caused by residual palindromic end sequences resulting from a single enzyme digestion.

With the GeneArt High-Order Genetic Assembly System, what are the requirements for stitching oligonucleotides used for insertion editing versus deletion editing?

Stitching oligonucleotides used for insertion editing must have a 30-nucleotide overlap with each adjacent fragment in addition to the insertion bases (for a total length of up to 80-mer, including up to 20 insertion bases). See manual for diagram. Note: This applies for a 2-fragment assembly and the insertion applies only to the internal junction. Use a 40-bp overlap (i.e., an 80-mer oligonucleotide) for the remaining seamless junctions.


Stitching oligonucleotides used for deletion editing must have a 40-nucleotide overlap with each adjacent fragment, annealing up to 6 nucleotides from the junction into each fragment, thus leaving up to 6 bp at the end of each fragment to be deleted during transformation-associated recombination. See manual for diagram. Note: This applies for a 2-fragment assembly and the deletion applies only to the internal junction. Use a 40-bp overlap (i.e., an 80-mer oligonucleotide) for the remaining seamless junctions.

For the GeneArt High-Order Genetic Assembly System, what enzyme do you recommend for amplifying my DNA fragments?

We recommend using our Platinum PCR Supermix High Fidelity enzyme (Cat. No. 12532-016) for amplifying DNA fragments up to 5 kb for general assembly applications. If you require higher PCR fidelity, we would recommend one that has processing and proof-reading capabilities.

I am using the GeneArt High-Order Genetic Assembly System and am trying to assemble large pieces of DNA. Do you have any recommendations?

Large fragments (>5 kb) are more susceptible to damage in a gel extraction procedure. Therefore, when using DNA fragments generated by PCR, we recommend that you assemble multiple fragments of less than 5 kb in one reaction rather than a single large fragment. Additionally, try to minimize UV exposure of the DNA and/or limit EtBr amount used. If you are attaching the insert to the linearized cloning vector, all nucleotides providing the requisite homology must be on the 5' end of the primer. If you are connecting two adjacent inserts, you may split the 30- to 50-bp homology between the fragments (e.g., 15 bp on the reverse primer of fragment 1 and 15 bp on the forward primer of fragment 2 for a 30-bp homology, or 25 bp on the reverse primer of fragment 1 and 25 bp on the forward primer of fragment 2 for a 50-bp homology). Please note, you can split the homology between adjacent fragments in any combination (e.g., 15 + 15 as in the example above or 20 + 10, 25 + 5 etc. for a 30-bp homology).

What are the specifications for the pYES1L vector? Can I use this as my cloning vector?

The pYES1L vector, a 9.4 kb linearized YAC-BAC shuttle plasmid, is a control vector for the assembly of DNA fragments in MaV203 cells and for the transfer of the assembled recombinant DNA molecule into E. coli. You can also use the pYES1L vector as a cloning vector, if desired. The origin of replication for yeast is chromosome II's centromere (single copy, high capacity YAC). The origin of replication for E. coli is F' ori (single copy, high capacity BAC). Spectinomycin is used for selection in E. coli. Four reactions (1 tube with 8µL) of the pYES1L vector are included with the kit. This is enough for control reactions, for cloning your inserts, or a combination of both. The pYES1L vector is also available separately (Cat. No. A13287, 10 reactions).

Will the addition of A-overhangs by the PCR enzyme affect cloning efficiency with the GeneArt High-Order Genetic Assembly System?

No, you can use enzymes that leave blunt ends or A-overhangs. Cloning efficiency will not be affected.

What cloning efficiency can be expected using the GeneArt High-Order Genetic Assembly System?

Cloning efficiencies can vary greatly based on the number of fragments with end-homology. Below are the approximate cloning efficiencies of assembled fragments into pYES1L:
>90% for 5 DNA fragments of 10 kb each
>90% for 10 DNA fragments of 5 kb each
>50% for 10 DNA fragments of 10 kb each
For pre-existing fragments without end-homology assembled into pYES1L using ‘stitching' DNA oligonucleotides, common cloning efficiencies are:
>90% for 1 fragment of 10 kb
>75% for 2 DNA fragments of 10 kb each

How do I use the web-based tool for fragment editing with the GeneArt High-Order Genetic Assembly System?

The web tool is not set up to do fragment editing automatically, but it can be used to do it manually. The manual has a section on fragment editing that gives examples on how to use the tool for this purpose.

I'm cloning 4 DNA fragments in a vector. Should I use the GeneArt Seamless Cloning and Assembly (Plus) Kit or the GeneArt High-Order Genetic Assembly Kit?

If the fragments are all below 5 kb and the total size of the molecule is below 13 kb, we would recommend the GeneArt Seamless Cloning and Assembly (Plus) kit. If you are assembling elements that have no end-homology, are too large to be amplified by PCR, or are trying to create a molecule over 13 kb, we recommend the GeneArt High-Order Genetic Assembly System. Overall, the High-Order system can do more fragments, fragment editing, and oligonucleotide stitching and can do it at a higher efficiency. However, the assembly is in vivo (yeast) and takes longer to get the final construct due to longer growth periods for yeast compared to E. coli. Select the GeneArt Seamless Cloning and Assembly kit that is best for your application by visiting http://www.thermofisher.com/us/en/home/life-science/cloning/seamless-cloning-and-genetic-assembly.html.

With the GeneArt High-Order Genetic Assembly System, can I assemble a molecule with fragments that carry homology (PCR) and others that are pre-existing (stitching) at the same time?

Yes, the kit can assemble up to 5 fragments and 3 oligo stitches.

Do you recommend a particular purity of oligonucleotides for the GeneArt High-Order Genetic Assembly System?

We have developed the kit using desalted oligonucleotides, but higher purities may result in slightly better cloning efficiencies, especially during fragment editing.

With the GeneArt High-Order Genetic Assembly System, how many overlapping nucleotides must I have if my construct is larger than 60 kb?

We recommend having a total overlap of 50 nucleotides with adjacent fragments for constructs larger than 60 kb. For constructs smaller than 60 kb, a total overlap of 30 nucleotides is recommended.


WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.