Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GeneArt™ Linear pUC19L Vector for Seamless Cloning

Catalog No. A13289
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13289 20 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A13289 Supplier Invitrogen™ Supplier No. A13289

The GeneArt™ Linear pUC19L Vector is a ready-to-use, linear version of the popular cloning vector pUC19.

The GeneArt™ Linear pUC19L Vector is a ready-to-use, linear version of the popular cloning vector pUC19. The pUC19L plasmid (2.7 Kbp), is specifically designed as a cloning and DNA assembly vector for the GeneArt™ Seamless Cloning and Assembly Kit (Cat. No. A13288). The vector is manufactured by digesting supercoiled pUC19 plasmid with the restriction enzymes PstI and KpnI, and tested for cloning of up to 4 DNA fragments.

Some of the key features and elements of the pUC19L vector include:

  • lacZα gene allows easy blue/white colony screening for selection of recombinants
  • pUC origin of replication (high copy vector)
  • Ampicillin resistance for selection in E. coli
  • 6 convenient and validated restriction sites flanking the cloning region
  • Tested for functionality by assembly with 4 DNA fragments using the GeneArt™ Seamless Cloning and Assembly Kit
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cloning Vector
Reporter Gene Beta-Gal (lacZ)
Content And Storage

Contains 5 vials of linear pUC19L vector at a concentration of 50 ng/mL. Each vial contains enough plasmid for 4 cloning reactions using the GeneArt™ Seamless Cloning and Assembly Kit (Cat. No. A13288, must be purchased separately).

Store the product at -80°C.
Vector is guaranteed stable for 6 months when properly stored.

Antibiotic Resistance Bacterial Ampicillin (AmpR)
Replication Origin pUC Origin
Shipping Condition Dry Ice
Size Range 13 kb total (vector plus all inserts)
Cloning Method Seamless Cloning
Concentration 50 ng/mL
Quantity 20 Reactions
Vector Seamless Cloning Vectors
Product Line GeneArt
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.