Learn More
Thermo Scientific™ GeneBLAzer™ ERRα-UAS-bla HEK 293T Cells
Description
ERR alpha-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Estrogen related receptor alpha (ERR alpha) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line. GeneBLAzer™UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-ERR alpha (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. ERR alpha-UAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of XCT790, an inverse agonist. In addition, ERR alpha-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.
Specifications
Specifications
| Alternate Agonists | L165, 041, GW 1929, GW 7647, GW 0742 |
| Antagonists | T0070907, GW 9662, GW 6471 |
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | GeneBLAzer™ Nuclear Receptor Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Detection Method | Fluorescence |
| Druggable Target | Nuclear Receptors |
| No. of Cells | 2 x 106 Cells |
| Primary Agonist | Rosiglitazone |
| Product Line | GeneBLAzer |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.