Learn More
GeneChip™ System 3000
Catalog No. 000218
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
00-021-8 1 Ea.
1 options
Catalog No. 00-021-8 Supplier LIFE TECHNOLOGIES Supplier No. 000218
This product is not available for sale online. Please contact Customer Service at 1-800-766-7000.
The GeneChip instrument system is a fully integrated platform for conducting your research using GeneChip brand probe arrays.
The GeneChip instrument system is a fully integrated platform for conducting your research using GeneChip brand probe arrays. The GeneChip Scanner 3000 7G System includes the GeneChip Scanner 3000 7G pre-assembled with the Autoloader, Fluidics Station 450, and Hybridization Oven 645i. This system can be purchased with or without a powerful computer workstation with quad-core Intel Core i9-12900 processor loaded with GeneChip Data Collection (GCDC) software. The instrument is used for multiple application areas, such as whole-genome association and targeted genotyping.
The purchase price includes a one-year service agreement.
Related Links:
*GeneChip Scanner 3000 High-Resolution Update is standard on all instruments shipped starting in September 2003 with serial number series 502. Previous versions, such as serial number series 501, will require the GeneChip Scanner 3000 High-Resolution Update, Catalog Number 00-0110, to be installed.
TRUSTED_SUSTAINABILITY
Specifications
Dimensions 22.5 x 31 x 44.5 in. (GeneChip Scanner 3000 with Autoloader)
28 x 16.1 x 15.8 in. (GeneChip Fluidic Station 450)
Product Line GeneChip
For Use With (Application) GeneChip Array Scanning
Weight 105 lb. (GeneChip Scanner 3000 with Autoloader), 80 lb. (GeneChip Fluidic Station 450)
Quantity 1 Ea.
For Research Use Only. Not for use in diagnostic procedures.